siRNA Details
| Gene Symbol: Rab15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134050.4 | NP_598811.3 | 3 | 416 | ||
| GenBank mRNA | AB232610.1 | - | 3 | 209 | ||
| AK158637.1 | - | 3 | 338 | |||
| AK158768.1 | - | 3 | 370 | |||
| BC013790.1 | - | 3 | 362 | 2655151 | ||
| BC027769.1 | - | 3 | 416 | 5345297 | ||

Rab15
Genomic Map