siRNA Details
| Gene Symbol: Epdr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134065.4 | NP_598826.3 | 2 | 723 | ||
| GenBank mRNA | AF353717.1 | - | 2 | 536 | ||
| AK034083.1 | - | 2 | 692 | |||
| AK038733.1 | - | 2 | 698 | |||
| AK046185.1 | - | 2 | 698 | |||
| AK160413.1 | - | 2 | 658 | |||
| AK166252.1 | - | 2 | 664 | |||
| BC014708.1 | - | 2 | 678 | 4223134 | ||

Epdr1
Genomic Map