siRNA Details
| Gene Symbol: Isy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133934.4 | NP_598695.1 | 5 | 289 | ||
| GenBank mRNA | AK017996.1 | - | 5 | 206 | ||
| AK173116.1 | - | 257 | ||||
| BC020103.1 | - | 5 | 253 | 3673446 | ||
| BC037695.1 | - | 5 | 294 | 4009615 | ||

Isy1
Genomic Map