siRNA Details
| Gene Symbol: Vrk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133945.1 | NP_598706.1 | 5, 6 | 563 | ||
| GenBank mRNA | AK150435.1 | - | 5, 6 | 654 | ||
| AK171673.1 | - | 5, 6 | 568 | |||
| BC010473.1 | - | 4, 5 | 546 | 3586304 | ||
| BC024782.1 | - | 4, 5 | 546 | 5123949 | ||
| BC024839.1 | - | 5, 6 | 563 | 5355521 | ||
| BC034196.1 | - | 4, 5 | 544 | 4985176 | ||

Vrk3
Genomic Map