siRNA Details
| Gene Symbol: Usb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133954.2 | NP_598715.2 | 3, 4 | 541 | ||
| GenBank mRNA | AK135527.1 | - | 3, 4 | 534 | ||
| AK160901.1 | - | 3, 4 | 524 | |||
| AK168683.1 | - | 3, 4 | 523 | |||
| AK182146.1 | - | 28 | ||||
| BC016418.1 | - | 3, 4 | 485 | 4224590 | ||

Usb1
Genomic Map