siRNA Details
| Gene Symbol: Rhou | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133955.4 | NP_598716.1 | 2, 3 | 355 | ||
| GenBank mRNA | AB051827.1 | - | 2, 3 | 355 | ||
| AF378088.1 | - | 2, 3 | 338 | |||
| BC134407.1 | - | 3, 4 | 192 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhou| Gene Symbol: Rhou | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133955.4 | NP_598716.1 | 2, 3 | 355 | ||
| GenBank mRNA | AB051827.1 | - | 2, 3 | 355 | ||
| AF378088.1 | - | 2, 3 | 338 | |||
| BC134407.1 | - | 3, 4 | 192 | |||