siRNA Details
| Gene Symbol: Trf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133977.2 | NP_598738.1 | 7 | 786 | ||
| GenBank mRNA | AF440692.1 | - | 6 | 716 | ||
| AK085754.1 | - | 7 | 771 | |||
| AK142599.1 | - | 776 | ||||
| AK146549.1 | - | 7 | 771 | |||
| AK149559.1 | - | 7 | 771 | |||
| AK149595.1 | - | 7 | 771 | |||
| AK150782.1 | - | 7 | 819 | |||
| AK168419.1 | - | 7 | 771 | |||
| AK168938.1 | - | 7 | 771 | |||
| AK169187.1 | - | 7 | 770 | |||
| BC008559.1 | - | 5 | 508 | |||
| BC012313.1 | - | 7 | 786 | 3981648 | ||
| BC018573.1 | - | 2 | 56 | |||
| BC020295.1 | - | 7 | 755 | |||
| BC022986.1 | - | 7 | 764 | 5123741 | ||
| BC058216.1 | - | 7 | 752 | |||
| BC058218.1 | - | 7 | 765 | |||
| BC092046.1 | - | 7 | 747 | 3594705 | ||

Trf
Genomic Map