siRNA Details
| Gene Symbol: Il13ra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080147.1 | - | 2, 3 | 256 | ||
| AK082889.1 | - | 2, 3 | 251 | |||
| AK089984.1 | - | 2, 3 | 287 | |||
| AK143292.1 | - | 2, 3 | 256 | |||
| AK154675.1 | - | 2, 3 | 311 | |||
| BC052425.1 | - | 2, 3 | 279 | 6835500 | ||
| BC059939.1 | - | 2, 3 | 247 | 6477224 | ||
| S80963.1 | - | 2, 3 | 284 | |||

Il13ra1
Genomic Map