siRNA Details
| Gene Symbol: Csnk1g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159591.1 | NP_001153063.1 | 8, 9 | 1179 | ||
| NM_134002.2 | NP_598763.1 | 7, 8 | 1356 | |||
| GenBank mRNA | AK077076.1 | - | 8, 9 | 1159 | ||
| AK132871.1 | - | 8, 9 | 1180 | |||
| BC004839.1 | - | 7, 8 | 1298 | 2648652 | ||
| BC063083.1 | - | 5, 6 | 2480 | |||

Csnk1g2
Genomic Map