siRNA Details
Gene Symbol: Akr1c18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134066.2 | NP_598827.1 | 4 | 414 | ||
GenBank mRNA | AB059565.1 | - | 4 | 414 | ||
AK153898.1 | - | 4 | 432 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Akr1c18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134066.2 | NP_598827.1 | 4 | 414 | ||
GenBank mRNA | AB059565.1 | - | 4 | 414 | ||
AK153898.1 | - | 4 | 432 |