siRNA Details
| Gene Symbol: Trf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133977.2 | NP_598738.1 | 13 | 1674 | ||
| GenBank mRNA | AF440692.1 | - | 12 | 1604 | ||
| AK085754.1 | - | 13 | 1659 | |||
| AK142599.1 | - | 1664 | ||||
| AK146549.1 | - | 13 | 1659 | |||
| AK149559.1 | - | 13 | 1659 | |||
| AK149595.1 | - | 13 | 1659 | |||
| AK150782.1 | - | 13 | 1707 | |||
| AK168368.1 | - | 13 | 1661 | |||
| AK168405.1 | - | 13 | 1661 | |||
| AK168419.1 | - | 13 | 1659 | |||
| AK168938.1 | - | 13 | 1659 | |||
| AK169187.1 | - | 13 | 1658 | |||
| AK203903.1 | - | 1 | 81 | |||
| BC008559.1 | - | 11 | 1396 | |||
| BC012313.1 | - | 13 | 1674 | 3981648 | ||
| BC018573.1 | - | 8 | 944 | |||
| BC020295.1 | - | 13 | 1643 | |||
| BC022986.1 | - | 13 | 1652 | 5123741 | ||
| BC058216.1 | - | 13 | 1640 | |||
| BC058218.1 | - | 13 | 1653 | |||
| BC092046.1 | - | 13 | 1635 | 3594705 | ||
| M23016.1 | - | 1 | 29 | |||

Trf
Genomic Map