siRNA Details
| Gene Symbol: C8G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000606.2 | NP_000597.2 | 5 | 611 | ||
| GenBank mRNA | BC113624.1 | - | 5 | 586 | 8322676 | |
| BC113626.1 | - | 5 | 586 | 8322678 | ||
| BX282392.1 | - | 4 | 349 | |||
| M17263.1 | - | 5 | 562 | |||
| M17999.1 | - | 5 | 597 | |||
| X06465.1 | - | 5 | 597 | |||

C8G
Genomic Map