siRNA Details
| Gene Symbol: Csnk1g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159591.1 | NP_001153063.1 | 12 | 1538 | ||
| NM_134002.2 | NP_598763.1 | 11 | 1715 | |||
| GenBank mRNA | AK077076.1 | - | 12 | 1518 | ||
| AK132871.1 | - | 12 | 1539 | |||
| AK152413.1 | - | 11 | 1714 | |||
| BC004839.1 | - | 11 | 1657 | 2648652 | ||
| BC005750.1 | - | 9 | 744 | 3600496 | ||
| BC063083.1 | - | 6 | 3436 | |||

Csnk1g2
Genomic Map