siRNA Details
| Gene Symbol: Mbtd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134012.3 | NP_598773.2 | 15, 16 | 2017 | ||
| GenBank mRNA | AK028503.1 | - | 8, 9 | 959 | ||
| AK153919.1 | - | 14, 15 | 1329 | |||
| AK161836.1 | - | 3, 4 | 400 | |||
| BC020018.1 | - | 11, 12 | 1328 | 2646754 | ||
| BC062907.1 | - | 15, 16 | 2017 | 6813849 | ||

Mbtd1
Genomic Map