siRNA Details
| Gene Symbol: Tbc1d10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134023.1 | NP_598784.1 | 7, 8 | 961 | ||
| GenBank mRNA | AK140872.1 | - | 7, 8 | 973 | ||
| AK153677.1 | - | 749 | ||||
| AK220243.1 | - | 1509 | ||||
| BC018300.1 | - | 7, 8 | 961 | 4193046 | ||
| BC029773.1 | - | 5, 6 | 525 | |||
| BC037230.1 | - | 7, 8 | 944 | 2650923 | ||

Tbc1d10a
Genomic Map