siRNA Details
| Gene Symbol: Hoxb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134032.2 | NP_598793.2 | 2 | 1484 | ||
| GenBank mRNA | BC019414.1 | - | 1 | 1069 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxb2| Gene Symbol: Hoxb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134032.2 | NP_598793.2 | 2 | 1484 | ||
| GenBank mRNA | BC019414.1 | - | 1 | 1069 | ||