siRNA Details
| Gene Symbol: Sptssa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134054.2 | NP_598815.2 | 2 | 953 | ||
| GenBank mRNA | AK003171.1 | - | 2 | 890 | ||
| AK008904.1 | - | 2 | 910 | |||
| AK018380.1 | - | 2 | 930 | |||
| AK137543.1 | - | 2 | 928 | |||
| BC022674.1 | - | 2 | 879 | 4236922 | ||

Sptssa
Genomic Map