siRNA Details
| Gene Symbol: Fam19a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252310.1 | NP_001239239.1 | 3 | 520 | ||
| NM_134096.2 | NP_598857.1 | 3 | 472 | |||
| GenBank mRNA | AK090194.1 | - | 3 | 346 | ||
| AK090307.1 | - | 3 | 522 | |||
| AY325124.1 | - | 3 | 473 | |||
| BB873716.1 | - | 2 | 182 | |||
| BC015306.1 | - | 3 | 473 | 4223181 | ||
| BI986174.1 | - | 2 | 152 | |||
| BU057253.1 | - | 3 | 478 | |||
| BY733542.1 | - | 3 | 522 | |||

Fam19a5
Genomic Map