siRNA Details
| Gene Symbol: Mfsd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134100.4 | NP_598861.1 | 2 | 422 | ||
| GenBank mRNA | AK087550.1 | - | 2 | 425 | ||
| AK089268.1 | - | 2 | 425 | |||
| AK165774.1 | - | 2 | 394 | |||
| BC009140.1 | - | 1 | 329 | 3598007 | ||

Mfsd5
Genomic Map