siRNA Details
| Gene Symbol: Wdr74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134139.1 | NP_598900.1 | 3 | 214 | ||
| GenBank mRNA | AK077521.1 | - | 3 | 215 | ||
| AK145704.1 | - | 3 | 227 | |||
| BC019968.1 | - | 3 | 214 | 4167260 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr74| Gene Symbol: Wdr74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134139.1 | NP_598900.1 | 3 | 214 | ||
| GenBank mRNA | AK077521.1 | - | 3 | 215 | ||
| AK145704.1 | - | 3 | 227 | |||
| BC019968.1 | - | 3 | 214 | 4167260 | ||