siRNA Details
| Gene Symbol: AI837181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256515.1 | NP_001243444.1 | 2 | 393 | ||
| NM_134149.2 | NP_598910.2 | 2 | 379 | |||
| GenBank mRNA | AK077296.1 | - | 1 | 115 | ||
| BC017542.1 | - | 2 | 260 | 4526963 | ||
| BC024516.1 | - | 2 | 380 | 4984074 | ||
| CJ181010.1 | - | 2 | 384 | |||
| CK622392.1 | - | 2 | 373 | |||
| U64690.1 | - | 2 | 258 | |||

AI837181
Genomic Map