siRNA Details
Gene Symbol: Vmn1r234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134198.1 | NP_598959.1 | 1 | 261 | ||
GenBank mRNA | BC127026.1 | - | 1 | 261 | 40044289 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vmn1r234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134198.1 | NP_598959.1 | 1 | 261 | ||
GenBank mRNA | BC127026.1 | - | 1 | 261 | 40044289 |