siRNA Details
| Gene Symbol: Kctd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111028.1 | NP_001104498.1 | 10 | 1135 | ||
| NM_001285933.1 | NP_001272862.1 | 10 | 1262 | |||
| NM_134073.2 | NP_598834.2 | 9 | 985 | |||
| GenBank mRNA | AK087312.1 | - | 10 | 1009 | ||
| AK158922.1 | - | 9 | 987 | |||
| AK166501.1 | - | 9 | 821 | |||
| BC011302.1 | - | 10 | 1076 | |||
| CK618608.1 | - | 3 | 280 | |||

Kctd9
Genomic Map