siRNA Details
| Gene Symbol: Cyp4f15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134127.1 | NP_598888.1 | 4 | 522 | ||
| GenBank mRNA | AF233645.1 | - | 4 | 522 | ||
| BC021377.1 | - | 4 | 513 | 5050617 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp4f15| Gene Symbol: Cyp4f15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134127.1 | NP_598888.1 | 4 | 522 | ||
| GenBank mRNA | AF233645.1 | - | 4 | 522 | ||
| BC021377.1 | - | 4 | 513 | 5050617 | ||