siRNA Details
| Gene Symbol: Smim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134133.2 | NP_598894.1 | 2 | 937 | ||
| GenBank mRNA | AF313412.1 | - | 2 | 479 | ||
| AK143383.1 | - | 2 | 941 | |||
| AK155873.1 | - | 2 | 583 | |||
| AK168283.1 | - | 2 | 760 | |||
| AK171810.1 | - | 2 | 595 | |||
| BC012898.1 | - | 2 | 445 | 4013357 | ||

Smim3
Genomic Map