siRNA Details
Gene Symbol: Vmn1r28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134180.1 | NP_598941.1 | 1 | 467 | ||
GenBank mRNA | AF394949.1 | - | 623 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vmn1r28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134180.1 | NP_598941.1 | 1 | 467 | ||
GenBank mRNA | AF394949.1 | - | 623 |