siRNA Details
| Gene Symbol: Vmn1r226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134191.1 | NP_598952.1 | 1 | 646 | ||
| GenBank mRNA | BC119429.1 | - | 1 | 644 | ||
| BC119430.1 | - | 1 | 646 | 40040474 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r226| Gene Symbol: Vmn1r226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134191.1 | NP_598952.1 | 1 | 646 | ||
| GenBank mRNA | BC119429.1 | - | 1 | 644 | ||
| BC119430.1 | - | 1 | 646 | 40040474 | ||