siRNA Details
| Gene Symbol: Chmp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134078.4 | NP_598839.2 | 10 | 2093 | ||
| GenBank mRNA | AK011732.1 | - | 1 | 474 | ||
| AK031882.1 | - | 1 | 394 | |||
| AK216196.1 | - | 1 | 52 | |||
| BC006648.1 | - | 4 | 2756 | |||
| BC024115.1 | - | 10 | 2081 | 5100379 | ||
| BC033365.1 | - | 8 | 2523 | 5367653 | ||

Chmp7
Genomic Map