siRNA Details
| Gene Symbol: Fam19a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252310.1 | NP_001239239.1 | 4 | 2555 | ||
| NM_134096.2 | NP_598857.1 | 4 | 2507 | |||
| GenBank mRNA | AK090307.1 | - | 4 | 2558 | ||
| BC015306.1 | - | 4 | 2508 | 4223181 | ||

Fam19a5
Genomic Map