siRNA Details
| Gene Symbol: Sft2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134114.2 | NP_598875.2 | 8 | 862 | ||
| GenBank mRNA | AK017471.1 | - | 6 | 606 | ||
| AK087027.1 | - | 3 | 405 | |||
| AK151338.1 | - | 8 | 793 | |||
| AK171776.1 | - | 6 | 627 | |||
| BC006777.1 | - | 7 | 2252 | |||
| BC091770.1 | - | 8 | 863 | 4974098 | ||

Sft2d1
Genomic Map