siRNA Details
| Gene Symbol: Trip10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242389.1 | NP_001229318.1 | 9 | 903 | ||
| NM_001242390.1 | NP_001229319.1 | 9 | 903 | |||
| NM_001242391.1 | NP_001229320.1 | 9 | 903 | |||
| NM_134125.4 | NP_598886.1 | 9 | 903 | |||
| GenBank mRNA | AF502565.1 | - | 9 | 806 | ||
| AK088909.1 | - | 9 | 853 | |||
| AK149902.1 | - | 9 | 875 | |||
| AY081142.1 | - | 9 | 854 | |||
| BC003249.1 | - | 9 | 905 | 3500118 | ||

Trip10
Genomic Map