siRNA Details
| Gene Symbol: AI837181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256515.1 | NP_001243444.1 | 2 | 1289 | ||
| NM_134149.2 | NP_598910.2 | 2 | 1275 | |||
| GenBank mRNA | AK077296.1 | - | 1 | 1011 | ||
| AV579065.1 | - | 1 | 192 | |||
| BC017542.1 | - | 2 | 1156 | 4526963 | ||
| BC024516.1 | - | 2 | 1276 | 4984074 | ||
| U64690.1 | - | 2 | 1154 | |||

AI837181
Genomic Map