siRNA Details
| Gene Symbol: Vmn1r38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134168.1 | NP_598929.1 | 1 | 786 | ||
| Gene Symbol: Vmn1r34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166719.1 | NP_001160191.1 | 1 | 786 | ||

Vmn1r38
Genomic Map