siRNA Details
| Gene Symbol: Vmn1r234 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134198.1 | NP_598959.1 | 1 | 715 | ||
| GenBank mRNA | BC127026.1 | - | 1 | 715 | 40044289 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r234| Gene Symbol: Vmn1r234 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134198.1 | NP_598959.1 | 1 | 715 | ||
| GenBank mRNA | BC127026.1 | - | 1 | 715 | 40044289 | |