siRNA Details
| Gene Symbol: Vmn1r212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134241.1 | NP_599002.1 | 1 | 115 | ||
| GenBank mRNA | BC132251.1 | - | 1 | 159 | 40130528 | |
| BC132253.1 | - | 1 | 159 | 40130530 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r212| Gene Symbol: Vmn1r212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134241.1 | NP_599002.1 | 1 | 115 | ||
| GenBank mRNA | BC132251.1 | - | 1 | 159 | 40130528 | |
| BC132253.1 | - | 1 | 159 | 40130530 | ||