siRNA Details
| Gene Symbol: Timd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161355.1 | NP_001154827.1 | 3 | 535 | ||
| NM_001161356.1 | NP_001154828.1 | 3 | 308 | |||
| NM_134249.5 | NP_599010.4 | 3 | 417 | |||
| GenBank mRNA | AB009015.1 | - | 3 | 308 | ||
| AF399827.1 | - | 3 | 269 | |||
| AF399828.1 | - | 3 | 269 | |||
| AK145630.1 | - | 3 | 296 | |||
| AK163225.1 | - | 3 | 296 | |||
| AK166877.1 | - | 3 | 289 | |||
| BC028829.1 | - | 3 | 379 | 5102129 | ||
| BY731803.1 | - | 3 | 417 | |||

Timd2
Genomic Map