siRNA Details
| Gene Symbol: Slc12a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083902.1 | NP_001077371.1 | 4 | 436 | ||
| NM_134251.2 | NP_599012.2 | 4 | 405 | |||
| GenBank mRNA | AF319951.1 | - | 4 | 399 | ||
| BC030926.1 | - | 4 | 436 | 4910321 | ||

Slc12a8
Genomic Map