siRNA Details
| Gene Symbol: Calml4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138304.2 | NP_612177.1 | 3 | 150 | ||
| GenBank mRNA | AK008039.1 | - | 3 | 136 | ||
| AK146656.1 | - | 3 | 123 | |||
| AY061807.1 | - | 3 | 150 | |||
| BC013541.1 | - | 3 | 128 | 4241711 | ||
| BC023475.1 | - | 3 | 115 | 5042406 | ||

Calml4
Genomic Map