siRNA Details
| Gene Symbol: Exosc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138586.3 | NP_613052.1 | 1, 2 | 239 | ||
| GenBank mRNA | AK006475.1 | - | 1, 2 | 192 | ||
| AK007372.1 | - | 1, 2 | 228 | |||
| AK137780.1 | - | 202 | ||||
| AK155228.1 | - | 1, 2 | 191 | |||
| BB856072.1 | - | 1, 2 | 241 | |||
| BC034358.1 | - | 1, 2 | 185 | 2651418 | ||
| BC092393.1 | - | 1, 2 | 180 | |||
| BY196593.1 | - | 1, 2 | 237 | |||
| BY198385.1 | - | 1, 2 | 237 | |||

Exosc5
Genomic Map