siRNA Details
| Gene Symbol: Ubfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138589.2 | NP_613055.2 | 2 | 519 | ||
| GenBank mRNA | AK011862.1 | - | 2 | 275 | ||
| AK017851.1 | - | 2 | 296 | |||
| AK033428.1 | - | 2 | 511 | |||
| AK077395.1 | - | 2 | 542 | |||
| AK135258.1 | - | 2 | 277 | |||
| AK140891.1 | - | 2 | 278 | |||
| BC011313.1 | - | 2 | 522 | 3499738 | ||

Ubfd1
Genomic Map