siRNA Details
| Gene Symbol: D6Wsu163e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138594.3 | NP_613060.1 | 2 | 184 | ||
| GenBank mRNA | AK037095.1 | - | 2 | 189 | ||
| AK042976.1 | - | 2 | 172 | |||
| AK043521.1 | - | 2 | 176 | |||
| AK045541.1 | - | 2 | 185 | |||
| AK046707.1 | - | 2 | 172 | |||
| AK137201.1 | - | 2 | 175 | |||
| BC016429.1 | - | 1 | 132 | 4486831 | ||

D6Wsu163e
Genomic Map