siRNA Details
| Gene Symbol: Med10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138596.2 | NP_613062.1 | 2 | 204 | ||
| GenBank mRNA | AK003862.1 | - | 2 | 198 | ||
| AK013992.1 | - | 2 | 208 | |||
| AK156498.1 | - | 2 | 213 | |||
| AK215905.1 | - | 2 | 52 | |||
| BC048638.1 | - | 2 | 197 | 6771841 | ||
| BC056438.1 | - | 2 | 193 | 6818638 | ||
| BC059231.1 | - | 1 | 1726 | |||
| BC089312.1 | - | 1 | 1749 | |||

Med10
Genomic Map