siRNA Details
| Gene Symbol: Vmn1r-ps103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134211.2 | NP_598972.2 | 1 | 134 | ||
| GenBank mRNA | BC110566.1 | - | 1 | 134 | ||
| BC110567.1 | - | 1 | 134 | |||
| BC127139.1 | - | 1 | 132 | |||
| BC127140.1 | - | 1 | 133 | |||

Vmn1r-ps103
Genomic Map