siRNA Details
| Gene Symbol: Vmn1r200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134212.1 | NP_598973.1 | 1 | 787 | ||
| GenBank mRNA | BC131902.1 | - | 1 | 869 | 40130179 | |
| BC131904.1 | - | 1 | 869 | 40130181 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r200| Gene Symbol: Vmn1r200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134212.1 | NP_598973.1 | 1 | 787 | ||
| GenBank mRNA | BC131902.1 | - | 1 | 869 | 40130179 | |
| BC131904.1 | - | 1 | 869 | 40130181 | ||