siRNA Details
| Gene Symbol: Vmn1r206 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134216.1 | NP_598977.1 | 1 | 787 | ||
| GenBank mRNA | BC132625.1 | - | 1 | 887 | 40130902 | |
| BC138067.1 | - | 1 | 887 | 8861086 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r206| Gene Symbol: Vmn1r206 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134216.1 | NP_598977.1 | 1 | 787 | ||
| GenBank mRNA | BC132625.1 | - | 1 | 887 | 40130902 | |
| BC138067.1 | - | 1 | 887 | 8861086 | ||