siRNA Details
| Gene Symbol: Vmn1r201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134221.1 | NP_598982.1 | 1 | 465 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r201| Gene Symbol: Vmn1r201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134221.1 | NP_598982.1 | 1 | 465 | ||