siRNA Details
| Gene Symbol: H1foo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138311.2 | NP_612184.1 | 2 | 186 | ||
| GenBank mRNA | AK135944.1 | - | 2 | 190 | ||
| AK162137.1 | - | 2 | 165 | |||
| AK162154.1 | - | 2 | 183 | |||
| AK165127.1 | - | 3 | 268 | |||
| AY007195.1 | - | 2 | 186 | |||
| BC137916.1 | - | 2 | 182 | 8860935 | ||
| BC145046.1 | - | 2 | 182 | |||

H1foo
Genomic Map