siRNA Details
| Gene Symbol: Nme7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293313.1 | NP_001280242.1 | 6 | 677 | ||
| NM_138314.4 | NP_612187.2 | 6 | 680 | |||
| NM_178071.6 | NP_835172.1 | 6 | 677 | |||
| GenBank mRNA | AF202048.1 | - | 5 | 581 | ||
| AK044962.1 | - | 6 | 651 | |||
| AK085222.1 | - | 6 | 625 | |||
| AK133221.1 | - | 1148 | ||||
| AK144915.1 | - | 6 | 681 | |||
| AK164652.1 | - | 6 | 625 | |||
| BC038021.1 | - | 7 | 707 | 3963844 | ||
| BC049225.1 | - | 5 | 608 | 3483725 | ||

Nme7
Genomic Map