siRNA Details
| Gene Symbol: Aldh7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127338.1 | NP_001120810.1 | 3, 4 | 468 | ||
| NM_138600.4 | NP_613066.2 | 1, 2 | 274 | |||
| GenBank mRNA | AK004991.1 | - | 3, 4 | 402 | ||
| AK145873.1 | - | 1, 2 | 217 | |||
| AK160117.1 | - | 1, 2 | 201 | |||
| AK169195.1 | - | 1, 2 | 217 | |||
| AK187582.1 | - | 17 | ||||
| BC012407.1 | - | 1, 2 | 236 | 4191228 | ||
| BY017676.1 | - | 1, 2 | 274 | |||

Aldh7a1
Genomic Map