siRNA Details
| Gene Symbol: Txlnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138628.3 | NP_619534.2 | 2 | 512 | ||
| GenBank mRNA | AF422244.1 | - | 2 | 449 | ||
| AF422245.1 | - | 2 | 449 | |||
| AK052324.1 | - | 2 | 476 | |||
| AK084579.1 | - | 2 | 512 | |||
| AK084639.1 | - | 2 | 513 | |||
| AK085904.1 | - | 2 | 406 | |||
| AK137442.1 | - | 2 | 427 | |||
| BC141023.1 | - | 2 | 425 | 9055635 | ||

Txlnb
Genomic Map